Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Supplier: Corning
3985 3643 3985BC 3540 3542 3544 3766 3762 3764 3768 3770
CORN3985EA 3630 SGD
CORN3985 CORN3643 CORN3985BC CORN3540 CORN3542 CORN3544 CORN3766 CORN3762 CORN3764 CORN3768 CORN3770
Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Microplates HTS Microplates
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk