Microplate, NBS, 384-well, low flange, black flat bottom, polystyrene, Corning®
Supplier: Corning
3654
CORN3654EA 0 SGD
CORN3654
Microplate, NBS, 384-well, low flange, black flat bottom, polystyrene, Corning®
Microplates HTS Microplates
Black wall plates have reduced autofluorescence, higher signal to background ratio and lower CV's.
  • Without lid
  • Non sterile
Nonbinding (NBS) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions.
Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise.