PCR plates, 384-well, Corning® Thermowell®
Supplier: Corning
3757
CORN3757EA 1100 SGD
CORN3757
PCR plates, 384-well, Corning® Thermowell®
Microplates PCR Plates
PP, non sterile. Compatible with a full range of PCR, sequencing and Real-Time PCR equipment.
Thermowell® GOLD 384-well PCR plates feature exceptional dimensional stability following thermocycling, and are fully compatible with automation, commonly used thermal cyclers, and Applied Biosystems® sequencing adapters.
Ordering information: Sealing tape and sealing mats sold separately.