Extractors for specific heavy solvents, Accelerated One-Step™, Modular, PYREX®
Supplier: Corning
3915-C
CORN3915-CEA 2770 SGD
CORN3915-C
Extractors for specific heavy solvents, Accelerated One-Step™, Modular, PYREX®
Extractors Liquid-Liquid Extractors
This revolutionary modular extractor-concentrator performs extraction, concentration and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.
Removable Snyder column sidearm allows easy cleaning. Extraction and concentration steps can be performed outside a hood, although the use of a ventilation system is recommended above or near the units in case of a drop in condenser water temperature or loss of condenser cold water supply.
Certifications: Meets EPA test requirements outlined by SW-846 method 3520 and WasteMore Product Information