Liquid/Liquid Extractor System, PYREX®
Supplier: Corning
3920-M 3920-MBO
CORN3920-MEA 1250 SGD
CORN3920-M CORN3920-MBO
Liquid/Liquid Extractor System, PYREX®
Extractors Liquid-Liquid Extractors
Made of PYREX® borosilicate glass. These continuous extractor systems are for use with solvents of heavier specific gravity than sample liquid being extracted.
  • Extraction can be performed outside the hood
  • Designed to meet EPA test method requirements outlined by SW-846, RCRA, and CLP
The sidearm is sealed to the extractor body just below the 45/50 [ST] outer joint. The sidearm is sealed higher on the body to reduce the chance of water spilling over while draining remaining solvent from the extractor body at the end of the extraction. Extraction can beMore Product Information