PYREX® Continuous Liquid/Liquid Extractor Body Assembly, Heavier Than Water, Modular
Supplier: Corning
3922-MBO 3922-MSAO
CORN3922-MBOEA 771 SGD
CORN3922-MBO CORN3922-MSAO
PYREX® Continuous Liquid/Liquid Extractor Body Assembly, Heavier Than Water, Modular
Extractors Liquid-Liquid Extractors
Removable sidearm and crossover tube reduce chance of contamination by allowing easy, thorough cleaning.
Parts are interchangeable with One-Step™ extractor/concentrators. Slightly taller form allows for longer solvent residence time and more efficient extractions. Capacity: 1000mL (34oz.).
Ordering information: Complete unit requires, the extractor body, a condenser, a crossover tube, a sidearm column and a receiver flask as well as small and large ball joint clamps and Viton O-rings.