96-Well Polypropylene Storage Blocks, Corning®
Supplier: Corning
3956 3957 3960 3961 3959 3958
CORN3956EA 577 SGD
CORN3956 CORN3957 CORN3960 CORN3961 CORN3959 CORN3958
96-Well Polypropylene Storage Blocks, Corning®
Microplates HTS Microplates
Plates are available in V-bottom and round-bottom formats to meet a wide variety of assay and storage needs.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
All sterile products are certified RNase- and DNase-free. Built to the industry standard footprint.
Ordering information: Storage mats and sealing tapes sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable