Corning® Direct Overhead Drive Center Cap Assembly for Corning® Reusable Spinner Flasks, Corning
Supplier: Corning
400649
CORN400649EA 3360 SGD
CORN400649
Corning® Direct Overhead Drive Center Cap Assembly for Corning® Reusable Spinner Flasks, Corning
Closures Bottle Closures
The 120 mm Center Cap Assembly for Corning® Spinner Flasks is designed for use with direct overhead drive motor.
These cap assemblies required optional 8 L, 15 L or 36 L paddle assemblies and can be used on all 4512 series vertical sidearm flasks.