Modified Impeller Assembly for Magnetically-Driven 3L Corning® Proculture® Spinner Flask
Supplier: Corning
402648
CORN402648EA 648 SGD
CORN402648
Modified Impeller Assembly for Magnetically-Driven 3L Corning® Proculture® Spinner Flask
Flasks Spinner Flasks
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 3L Proculture® Spinner Flasks
  • Narrower impeller paddle design allows use with probes to create a small bioreactor
  • Non sterile
  • Reusable