Sterile containers, 250 ml, Corning®
Supplier: Corning
430179 430180 430181
CORN430179EA 398 SGD
CORN430179 CORN430180 CORN430181
Sterile containers, 250 ml, Corning®
Sample Containers General Purpose Containers
PP Container, transparent, with snap-on HDPE lid.
  • Graduated in both millilitres and ounces
  • Certified non pyrogenic
  • sterile
Serves as a beaker or storage container.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa