Cryogenic vial rack, Corning®
Supplier: Corning
431131
CORN431131EA 123 SGD
CORN431131
Cryogenic vial rack, Corning®
Racks Cryogenic Vial Racks
This reusable PP rack holds 50 cryogenic vials.
  • Locking feature for use with all Corning® self-standing vials
  • Allows one handed opening
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa