Polystyrene roller bottle with easy grip cap, Corning®
Supplier: Corning
431644
CORN431644EA 2770 SGD
CORN431644
Polystyrene roller bottle with easy grip cap, Corning®
Flasks Cell Culture Flasks
The bottle is made with one piece seamless construction, has a wide mouth and printed graduations.
The bottle is manufactured from virgin polystyrene and the cap is made of high density polyethylene. Both resins meet the USP Class VI requirements for plastic containers and closures.
This product is sterilised by gamma radiation and is certified nonpyrogenic. It has printed lot numbers to aid in product traceability.
To be used with USP Class VI certified C-FLEX® tubing set.
The orange easy grip cap features large knurls designed for ergonomic handling. The bottle is not tissue-More Product Information