All-day cooling and freezing workstations, Corning® CoolBox™ XT
Supplier: Corning
432022 432023 432026
CORN432022EA 1350 SGD
CORN432022 CORN432023 CORN432026
All-day cooling and freezing workstations, Corning® CoolBox™ XT
Coolers Benchtop Coolers
Ideal for transfering samples between labs, or in the field.
  • Over 16 hours of 0,5 to 4 °C cooling with lid closed and over 10 hours with the lid open
  • Reduces contamination by eliminating ice
  • Easy to clean with lab detergents or bleach
  • Patent-pending dual phase conductive XT Cooling core ensures stable and consistent sample cooling
  • Portable XT CoolBox systems can be used when a brief transfer of temperature sensitive samples to a robotic device is needed
  • May be used with dry ice in place of the Cooling Core to create a portable snap-freezing workstation
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable