Cryogenic vial grippers, Corning®
Supplier: Corning
432136
CORN432136EA 166 SGD
CORN432136
Cryogenic vial grippers, Corning®
Forceps
These cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials.
They enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen.