Multi-position magnetic stirrers, 4-position, Corning®
Supplier: Corning
440814
CORN440814EA 3480 SGD
CORN440814
Multi-position magnetic stirrers, 4-position, Corning®
Stirrers Magnetic Stirrers, Multi-position
This Corning® four position slow-speed stirrer has integral controls that can accommodate up to four one-litre vessels.
  • Compact design saves valuable bench space
  • Offers powerful stirring from 10 to 150 min⁻¹
  • Rugged lightweight glass filled resin housing
  • Maximum vessel diameter is 75 mm
This is designed to be use with Corning® glass and plastic spinner flasks to provide identical stirring speeds for both good mixing and high cell viability.
All four positions operate at the same speed. It operates on 120 V/60 Hz and is UL approved. A compact, lowMore Product Information