96 Well Cluster Tubes, Corning®
Supplier: Corning
4401 4408 4410 4411 4412 4413 4418
CORN4401EA 106 SGD
CORN4401 CORN4408 CORN4410 CORN4411 CORN4412 CORN4413 CORN4418
96 Well Cluster Tubes, Corning®
Tubes Culture Tubes
PP tubes, PE tube caps.
  • Composed of 96 PP tubes in a standard microplate format
  • 1,2 ml tubes available individually or in strips of 8 tubes
  • PE tube caps available in 8-cap strips
Packaging: Supplied bagged or racked.
Order Now
SPECIFICATIONS
Environmentally Preferable