Fernbach-style culture flask with baffles 2800 ml, PYREX®
Supplier: Corning
4423
CORN4423EA 525 SGD
CORN4423
Fernbach-style culture flask with baffles 2800 ml, PYREX®
Tubes Culture Tubes
This 2800 ml wide mouth PYREX® Fernbach-style culture flask is designed for culturing organisms requiring a large surface area to volume ratio. It can also be used in serum production. Triple baffled on the bottom outside edges to achieve maximal oxygen transfer to culture medium. Beaded neck rim is approximately 60 mm I.D. and accepts cotton plugs or No. 13 rubber stoppers. This flask has a large marking spot.
Caution: NOTE: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.