Erlenmeyer flask, delong neck, with extra deep baffles, PYREX®
Supplier: Corning
4446-2L 4446-1L 4446-500
CORN4446-2LEA 250 SGD
CORN4446-2L CORN4446-1L CORN4446-500
Erlenmeyer flask, delong neck, with extra deep baffles, PYREX®
Flasks Erlenmeyer Flasks
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These PYREX® Delong style shaker flasks have three extra deep baffles designed to provide greater agitation of solutions to improve oxygen or gas transfer when used with rotary or reciprocating shakers. The long neck reduces splashing and is designed for two-position polypropylene closures. Their uniform wall thickness provides the proper balance between mechanical strength andMore Product Information