DeLong shaker Erlenmeyer flask with extra deep baffles, PYREX®
Supplier: Corning
4446-250
CORN4446-250EA 394 SGD
CORN4446-250
DeLong shaker Erlenmeyer flask with extra deep baffles, PYREX®
Flasks Erlenmeyer Flasks
These PYREX® Delong style shaker flasks have three extra deep baffles designed to provide greater agitation of solutions to improve oxygen or gas transfer when used with rotary or reciprocating shakers.
The long neck reduces splashing and is designed for two-position polypropylene closures. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance. For convenience, these flasks include a marking spot. The approximate neck O.D. is 18 mm. For closures see Nos. 15551-252.
Ordering information: Accessories not included.