Pipette tips, universal, Corning®
Supplier: Corning
4803 4804 4806 4844 4845 4846 4860 4862 4863 4865 4866 4867 4868 9032 4713 4710 4711 4712 4714 4715
CORN4803EA 413 SGD
CORN4803 CORN4804 CORN4806 CORN4844 CORN4845 CORN4846 CORN4860 CORN4862 CORN4863 CORN4865 CORN4866 CORN4867 CORN4868 CORN9032 CORN4713 CORN4710 CORN4711 CORN4712 CORN4714 CORN4715
Pipette tips, universal, Corning®
Pipette Tips
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.