Condensers, low actinic, PYREX®
Supplier: Corning
52300-650
CORN52300-650EA 87.5 SGD
CORN52300-650
Condensers, low actinic, PYREX®
Condensers
This PYREX® air condenser is made from low actinic glass.
  • Air Reflux style condenser
  • Has an inner 24/40 standard taper joint with a drip tip
To protect light sensitive materials and is useful for condensing materials with boiling points above 150 °C.