Kjeldahl-flask, with standard ground joint, tooled long neck, PYREX®
Supplier: Corning
5420-100 5420-300 5420-500 5420-800
CORN5420-100EA 21.7 SGD
CORN5420-100 CORN5420-300 CORN5420-500 CORN5420-800
Kjeldahl-flask, with standard ground joint, tooled long neck, PYREX®
Flasks Kjeldahl Flasks
PYREX® borosilicate glass, long neck.
  • Have tooled finished necks for extra strength and uniform stopper fit
  • Optimum balance between thermal shock and chemical resistance
  • Can be used in ASTM E-258 procedure
Suitable for the determination of nitrogen. Care must be taken to ensure the bottoms of the flasks do not come in contact with the heating elements. Kjeldahl flask have ring supports for this purpose.
Order Now
SPECIFICATIONS
Environmentally Preferable