Erlenmeyer flasks, narrow neck, low actinic, with standard ground joint stopper, PYREX®
Supplier: Corning
55020-250
CORN55020-250EA 82.4 SGD
CORN55020-250
Erlenmeyer flasks, narrow neck, low actinic, with standard ground joint stopper, PYREX®
Flasks Erlenmeyer Flasks
PYREX® low actinic glass.
  • Very good chemical resistance
  • High mechanical strength
These narrow mouth have standard taper stoppers for secure sealing. These low actinic stained glass provides protection for materials sensitive to light. For convenience, these flasks are graduated to show approximate capacity.
Order Now
SPECIFICATIONS
Environmentally Preferable