Volumetric flasks, class A, low actinic, PYREX® EZ Access™
Supplier: Corning
55635-100 55635-25 55635-250 55635-50
CORN55635-100EA 602 SGD
CORN55635-100 CORN55635-25 CORN55635-250 CORN55635-50
Volumetric flasks, class A, low actinic, PYREX® EZ Access™
Flasks Volumetric Flasks
PYREX® glass, low actinic, with stopper.
  • Graduation line is sharp and permanent
  • Low actinic glass provides protection for materials sensitive to light
  • Chemical and thermal resistance
Ideal for microchemical work. Features wide mouth facilitates pipette filling and mixing.
The flasks are supplied with larger interchangeable glass stoppers.