Volumetric flasks, class A, with stopper, PYREX®
Supplier: Corning
5680-10 5680-100 5680-1L 5680-200 5680-25 5680-250 5680-2L 5680-50 5680-500
CORN5680-10EA 65.9 SGD
CORN5680-10 CORN5680-100 CORN5680-1L CORN5680-200 CORN5680-25 CORN5680-250 CORN5680-2L CORN5680-50 CORN5680-500
Volumetric flasks, class A, with stopper, PYREX®
Flasks Volumetric Flasks
PYREX® borosilicate glass.
Flasks feature sharp, permanent graduation line and large white block letter inscriptions. Body is machine-blown with sealed heavy-tubing neck for increased strength. Hollow glass ground NS stoppers, where included, are interchangeable.
All flasks are graduated to Class A volumetric tolerances and calibrated to meet ASTM specifications E-694, E-542, and E-288. Certified flasks are individually serialised and supplied with NIST-traceable certificate of identification and capacity.