Volumetric flasks for Bates sugar analysis, class A, with tooled rim, PYREX®
Supplier: Corning
5720-100
CORN5720-100EA 51 SGD
CORN5720-100
Volumetric flasks for Bates sugar analysis, class A, with tooled rim, PYREX®
Flasks Volumetric Flasks
PYREX® borosilicate glass, class A.
These flasks have short necks and slightly flared tops. It is used for the preparation of sugar solutions for polarisation tests.
Certifications: ASTM E-694.