Volumetric flasks, class A, glass, clear, NS glass stopper, PYREX®
Supplier: Corning
5820-100 5820-1L 5820-250 5820-2L
CORN5820-100EA 1390 SGD
CORN5820-100 CORN5820-1L CORN5820-250 CORN5820-2L
Volumetric flasks, class A, glass, clear, NS glass stopper, PYREX®
Flasks Volumetric Flasks
Made of PYREX® borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
Reusable volumetric flasks with glass stoppers. This flasks have an upper bulb with the one-tenth of flask rated capacity for the mixing of liquids or powders when shaken.
Certifications: ASTM E-694, E-542, E-288.
Order Now
SPECIFICATIONS
Environmentally Preferable