Volumetric flasks, class A, glass, clear, viscosimeter, PYREX®
Supplier: Corning
5860-60
CORN5860-60EA 60 SGD
CORN5860-60
Volumetric flasks, class A, glass, clear, viscosimeter, PYREX®
Flasks Volumetric Flasks
Rugged mold-blown construction.
Certifications: Calibrated in accordance with ASTM D-88 and E-102.