Aspirator bottles with protective coating and tubing outlet, PYREXPLUS®
Supplier: Corning
61220-1L 61220-250 61220-2L 61220-3X 61220-4L 61220-500
CORN61220-1LEA 647 SGD
CORN61220-1L CORN61220-250 CORN61220-2L CORN61220-3X CORN61220-4L CORN61220-500
Aspirator bottles with protective coating and tubing outlet, PYREXPLUS®
Bottles Aspirators
PYREXPLUS®, coated.
These aspirator bottles have a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and is autoclavable at 121 °C and resistant to thermal shock. Bottles have a tubular sidearm outlet (10 mm Ø) near the bottom for attachment of flexible plastic tubing and are very useful as a delivery/storage container for solutions. The tubulation is not coated to allow easy connection of standard size tubing. These bottles feature sturdy ring necks with a heavy duty rims and use No. 2 rubber stoppers.
Warning: Do notMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable