Lid for 96-well PCR plates, Corning® Thermowell®
Supplier: Corning
6520
CORN6520EA 217 SGD
CORN6520
Lid for 96-well PCR plates, Corning® Thermowell®
Lids Microplate Lids
Polycarbonate lids are thermally stable up to 135 °C and can be used on Corning® Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays.
Lids are intended to be covers only and do not prevent evaporation.