Hot plates, Corning® Scholar™ PC-170
Supplier: Corning
6795-170
CORN6795-170EA 391 SGD
CORN6795-170
Hot plates, Corning® Scholar™ PC-170
Heaters Hotplates
Economical hot plate with white enamel steel-top plate
Provides heat transfer up to 360°C (680°F). Dual heat shields dissipate heat and keep the case cool to the touch. Compact size saves on bench space.
Ordering information: Manufacturer's one-year warranty on parts and labor.