Magnetic stirrer, Scholar™ PC-171 Corning®
Supplier: Corning
6795-171
CORN6795-171EA 436 SGD
CORN6795-171
Magnetic stirrer, Scholar™ PC-171 Corning®
Stirrers Magnetic Stirrers
Designed with a 12,7×12,7cm (5×5") ABS plastic top, the Scholar™ PC-171 Stirrer is compact and provides vigorous stirring with an alternating current motor for long life.
  • Speed range: 100 to 1600 rpm
Units feature lightweight yet sturdy construction to allow rugged use. Space saving design allows users to maximise bench space.