High volume stirrer, Corning®
Supplier: Corning
6795-611
CORN6795-611EA 1680 SGD
CORN6795-611
High volume stirrer, Corning®
Stirrers Magnetic Stirrers
Ideal for stirring large volumes of liquid.
Designed for 19 L (5 gal.) vessels, but can stir up to 38 L (10 gal.). Compact design conserves bench space. Stirrer uses a magnet made of rare earth metals for exceptional coupling strength that helps center the stir bar quickly inside the vessel. Heavy-duty motor and rugged ABS plastic top make unit long-lasting. Top size is 279x279 mm (11x11").
Ordering information: Fluoropolymer resin-coated stir bar available separately.