Accessory Kit for Digital Display Hotplates and Hotplate Stirrers, Corning®
Supplier: Corning
6795KIT
CORN6795KITEA 451 SGD
CORN6795KIT
Accessory Kit for Digital Display Hotplates and Hotplate Stirrers, Corning®
Stirrers Magnetic Hotplate Stirrers
This accessory kit is designed for use with all Corning® digital display hotplates and stirring hotplates (Models PC-400D, PC-420D, PC-600D, PC-620D)
It is voltage independent. Each kit includes a temperature controller that controls the temperature of liquids up to 200 °C, a two-piece, 0.8 cm diameter, 45.7 cm long support rod, and magnetic stir bar retriever.