Hotplate, Corning®
Supplier: Corning
6796-200
CORN6796-200EA 856 SGD
CORN6796-200
Hotplate, Corning®
Heaters Hotplates
Low-profile hot plates with a ceramic glass plate provide precision-controlled heating for laboratory applications. Microprocessor-controlled heating ensures consistent performance.
  • Space saving design
  • Hot top indicator light
  • Large knobs provide control for gloved fingers
Units feature large knobs and a mount for a ring stand support built into the back of the housing. A hot top indicator light blinks a warning when the top is too hot to touch, >60 °C (140 °F). All units feature a temperature range from 25 to 550 °C (77 to 1022 °F).
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Model PC-200
Weight 1,8 kg
Environmental certification I-RECs, ISO 14001 - Reynosa