Magnetic stirring bar retrievers, Corning®
Supplier: Corning
6970SR
CORN6970SREA 19.5 SGD
CORN6970SR
Magnetic stirring bar retrievers, Corning®
Stirrers Stirring Tools
The stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory.