Volumetric flask, class B, blue inscriptions, PYREX® , VISTA™
Supplier: Corning
70581-10 70581-100 70581-1L 70581-25 70581-250 70581-50 70581-500
CORN70581-10EA 376 SGD
CORN70581-10 CORN70581-100 CORN70581-1L CORN70581-25 CORN70581-250 CORN70581-50 CORN70581-500
Volumetric flask, class B, blue inscriptions, PYREX® , VISTA™
Flasks Volumetric Flasks
Made of PYREX® glass, with PE snap-cap.
  • Heavy duty necks
  • Graduation line is sharp and permanent
  • Inscriptions easy to read
Designed for use by customers that do not require the precision accuracy of our Class A capacity flasks. The shape, body and wall are the same as regular flasks but display the large blue marking spot and designation 'PYREX VISTA' unique to the PYREX VISTA line. Capacity tolerances are twice those specified for Class A volumetric ware.
Heavy-duty necks are tooled for HDPE snap-caps, which are included with all sizes. Flat bottomMore Product Information