Culture tubes, rimless, PYREX® VISTA™
Supplier: Corning
70820-10 70820-12 70820-13 70820-16 70820-16X 70820-16XX 70820-18 70820-20 70820-22 70820-25 70820-25X 70820-6
CORN70820-10EA 80.3 SGD
CORN70820-10 CORN70820-12 CORN70820-13 CORN70820-16 CORN70820-16X CORN70820-16XX CORN70820-18 CORN70820-20 CORN70820-22 CORN70820-25 CORN70820-25X CORN70820-6
Culture tubes, rimless, PYREX® VISTA™
Tubes Culture Tubes
Borosilicate glass.
Reusable rimless culture tubes offer greater convenience in plugging and rack storage. Round bottom tubes feature fire polished ends. All tubes have a large blue enamel marking spot for simple identification, with the exception of 50, 55 and 70 ml sizes.
Delivery information: Supplied without caps.
Order Now
SPECIFICATIONS
Environmentally Preferable