Tissue grinders, Tenbroeck type, PYREX®
Supplier: Corning
7726-L
CORN7726-LEA 313 SGD
CORN7726-L
Tissue grinders, Tenbroeck type, PYREX®
Homogenisers Tissue Grinders
These homogenisers are designed for grinding small samples for pathological and biochemical work. The catch basin bulb has a 55 mm outer diameter.
Can be used manually but is adaptable for use with a slow speed stirrer by means of a rubber stopper and a connecting rod.
Certifications: Made in accordance with Federal Specification DD-T-363a.
Caution: If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus. The user should wear a protective glove of sufficient thickness to prevent injury in the event ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable