Drying tubes with standard taper joints, PYREX®
Supplier: Corning
7800-24
CORN7800-24EA 1480 SGD
CORN7800-24
Drying tubes with standard taper joints, PYREX®
Tubes Drying Tubes
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These inverted drying tubes have an inner 24/40 Standard Taper joint at one end only and a single bulb. The chamber is approximately 110 mm long including the bulb (30 mm Ext.Ø) and will take a No. 2 rubber stopper.
Order Now
SPECIFICATIONS
Environmentally Preferable