Digestion tubes, Folin-Wu, PYREX®
Supplier: Corning
7900-25
CORN7900-25EA 21.4 SGD
CORN7900-25
Digestion tubes, Folin-Wu, PYREX®
Tubes Digestion Tubes
Made of PYREX® borosilicate glass.
  • Round bottom
  • With engraved white-filled graduations at 25 and 50 ml
  • Tolerance of ±0,4 ml
The Folin-Wu digestion tubes are graduated with a beaded rim and are fabricated from specially selected tubing for product durability.