Conical centrifuge tubes with white graduations and standard taper pennyhead stopper, PYREX®
Supplier: Corning
8084-15
CORN8084-15EA 714 SGD
CORN8084-15
Conical centrifuge tubes with white graduations and standard taper pennyhead stopper, PYREX®
Tubes Centrifuge Tubes
PYREX®, borosilicate glass.
These tubes have white enamel graduations. The 5 and 15 ml sizes are graduated upward from the tip in 0,1 ml increments. The 50 ml size is graduated upward from tip to 10 ml in 0,5 ml increments, and from 10 ml to 50 in 1 ml increments. The pennyhead stopper is solid.
For stopper only, see Cat. No. 7661.