Conical centrifuge tubes with black graduations, beaded rim, PYREX®
Supplier: Corning
8101-15 8101-50
CORN8101-15EA 338 SGD
CORN8101-15 CORN8101-50
Conical centrifuge tubes with black graduations, beaded rim, PYREX®
Tubes Centrifuge Tubes
These tubes have sharp, coloured enamel graduations and markings.
The tubes are the same size and shape as Cat. Nos. 8060 and 8080. They have precision formed tips and uniform walls throughout for a high degree of accuracy and mechanical strength.
Order Now
SPECIFICATIONS
Environmentally Preferable