Centrifuge tube, Goetz, pear shape, PYREX®
Supplier: Corning
8220-100
CORN8220-100EA 1890 SGD
CORN8220-100
Centrifuge tube, Goetz, pear shape, PYREX®
Tubes Centrifuge Tubes
Made of PYREX® borosilicate glass.
  • With white graduations
  • Narrow mouth neck style
  • Round bottom
  • Beaded rim
This pear-shaped centrifuge tubes are used for phosphorus determinations in iron and steel.
The lower stem is graduated upward to 1 ml in 0,05 ml divisions. The body is graduated from 1 to 5 ml in 1 ml divisions; from 5 to 50 ml in 5 ml divisions; and from 50 to 100 ml in 10 ml divisions.