Centrifuge tubes, Kolmer, PYREX®
Supplier: Corning
8360-10
CORN8360-10EA 883 SGD
CORN8360-10
Centrifuge tubes, Kolmer, PYREX®
Tubes Centrifuge Tubes
Made of PYREX®, borosilicate glass.
  • Narrow neck and round bottom
  • With white enamel graduations
These tubes are used in connection with the Wasserman test and to measure water content and potassium ions in oil muds.
Graduated in 0,1 ml divisions up to 4 ml and 0,2 ml divisions from 4 to 10 ml.