Centrifuge tube, round bottom, PYREX®
Supplier: Corning
8424-50
CORN8424-50EA 450 SGD
CORN8424-50
Centrifuge tube, round bottom, PYREX®
Tubes Centrifuge Tubes
PYREX®, borosilicate glass, round bottom, with ground joint.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These PYREX® round bottom, non-graduated centrifuge tubes have a No. 13 standard taper solid glass pennyhead stopper.Rockefeller Institute of Medical Research type.
Reusable and non-sterile.