Gas chromatography collection tube, PYREX®
Supplier: Corning
9080
CORN9080EA 105 SGD
CORN9080
Gas chromatography collection tube, PYREX®
Tubes Gas Sampling Tubes
This gas chromatograph collection tube is a replacement part for the 6949M series of PYREX® microchemistry kits.