Standard polystyrene microplates 96-well, Corning® Stripwell™
Supplier: Corning
2580 2592 2593 3923 3924 9102
CORN2580EA 445 SGD
CORN2580 CORN2592 CORN2593 CORN3923 CORN3924 CORN9102
Standard polystyrene microplates 96-well, Corning® Stripwell™
Microplates Immunology Microplates and Strips
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an 'egg-crate' style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • 1×8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk