Snap caps for centrifuge tubes, Corning®
Supplier: Corning
99500-1
CORN99500-1EA 31.9 SGD
CORN99500-1
Snap caps for centrifuge tubes, Corning®
Closures Test/Sample Tube Closures
HDPE, orange.
These snap caps are manufactured to rigorous standards to ensure a leak proof seal. They are designed to fit PYREX® 5ml disposable glass centrifuge tubes.