Disposable glass conical centrifuge tubes, ungraduated, without screw cap, PYREX®
Supplier: Corning
99502-50
CORN99502-50EA 368 SGD
CORN99502-50
Disposable glass conical centrifuge tubes, ungraduated, without screw cap, PYREX®
Tubes Centrifuge Tubes
Corning disposable glass centrifuge tubes are manufactured from PYREX® Type I borosilicate glass.
These tubes feature more consistent uniform wall thickness, a well-formed conical tapered tip and durable tip radius. The optimally shaped tip design allows for easy removal of samples following centrifugation. The tubes are designed to withstand centrifugation up to 3000 RCF and are packed in convenient, easy to use packs, which are shrink sealed for protection during shipment.
Ordering information: All caps must be ordered separately. For compatible screw caps, select either disposableMore Product Information